SSPE Buffer, 20X, Liquid Concentrate, OmniPur*.
Supplier: MilliporeSigma
8410-4L
EM-8410EA 364.85 USD
EM-8410
SSPE Buffer, 20X, Liquid Concentrate, OmniPur*.
Buffers Electrophoresis Buffers Hybridization Buffers
Each liter of concentrate makes 20L of 1X SSPE (0.150M NaCl, 0.010M Sodium Phosphate, 0.001M EDTA).