SSC Buffer, Concentrate, OmniPur*. 20X.
Supplier: MilliporeSigma
8310-4L
EM-8310EA 293.86 USD
EM-8310
SSC Buffer, Concentrate, OmniPur*. 20X.
Buffers Electrophoresis Buffers Hybridization Buffers
Liquid
Each liter of concentrate makes 20L of 1X SSC (0.15M NaCl, 0.15M sodium citrate). DNase-, RNase-, and protease-free.