Sodium dodecyl sulfate (SDS) 20% in aqueous solution, OmniPur®, Millipore®

Supplier: MilliporeSigma

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

7990-200ML 7991-1L
EM-7990EA 135.05 USD
EM-7990 EM-7991
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, OmniPur®, Millipore®
Sodium dodecyl sulphate (SDS)

Clear, colorless liquid. For molecular biology. DNase-, RNase-, and protease-free.

Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Max. UV Absorbance at 260 nm 0.1
Max. UV Absorbance at 280 nm 0.1
Conductivity 21,900–25,500 µmhos
pH (at 25°C) 5.0–8.0
Phosphate (PO4) 0.0005% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR