Sodium dodecyl sulfate (SDS) 20% in aqueous solution, OmniPur®, Millipore®
Supplier: MilliporeSigma
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
7990-200ML
7991-1L
EM-7990EA
135.05
USD
EM-7990
EM-7991
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, OmniPur®, Millipore®
Sodium dodecyl sulphate (SDS)
Clear, colorless liquid. For molecular biology. DNase-, RNase-, and protease-free.
Formula:
CH₃(CH₂)₁₁OSO₃Na Density: 1…1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 |
|
Specification Test Results
Max. UV Absorbance at 260 nm | 0.1 |
Max. UV Absorbance at 280 nm | 0.1 |
Conductivity | 21,900–25,500 µmhos |
pH (at 25°C) | 5.0–8.0 |
Phosphate (PO4) | 0.0005% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...