Sodium dodecyl sulfate (SDS) ≥99.0%, OmniPur®, Millipore®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

7910-500GM 7960-5KG
EM-7910EA 483.93 USD
EM-7910 EM-7960
Sodium dodecyl sulfate (SDS) ≥99.0%, OmniPur®, Millipore®
Sodium dodecyl sulphate (SDS)

DNase-, RNase-, protease-, and phosphatase-free.

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR