Phosphate Buffered Saline (PBS) Tablets, OmniPur®, Calbiochem®
Supplier: MilliporeSigma
6501-200TAB
EM-6501EA 541.15 USD
EM-6501
Phosphate Buffered Saline (PBS) Tablets, OmniPur®, Calbiochem®
Buffers Biological Buffers
White, concave tablets
Each tablet makes 1X solution when dissolved in 100mL distilled or deionized water (1X=137mM sodium chloride, 2.7mM potassium chloride, and 10mM sodium phosphate dibasic/potassium phosphate monobasic buffer). If higher concentrations are desired, dissolve up to four tablets per 100mL water. DNase-, RNase-, and protease-free. Toxic effects.