γ-(N-Morpholino)propanesulfonic acid (MOPS) concentrate, OmniPur® 10X, Millipore®

Supplier: MilliporeSigma

6350-500ML
EM-6350EA 294.99 USD
EM-6350
γ-(N-Morpholino)propanesulfonic acid (MOPS) concentrate, OmniPur® 10X, Millipore®
γ-(N-Morpholino)propanesulphonic acid (MOPS)

Liquid. Prepared with MOPS and DEPC for RNA applications. Each liter of concentrate makes 10L of 1X MOPS. DNase-, RNase-, and protease-free.

Formula: C₇H₁₅NO₄S
MDL Number: MFCD00006183
CAS Number: 1132-61-2

Order Now

Specification Test Results

Expiration date 60 months
Titer (20 degrees C) 0.998-1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR