Lithium chloride 8 M, OmniPur®

Supplier: MilliporeSigma

5922-500ML
EM-5922EA 285.45 USD
EM-5922
Lithium chloride 8 M, OmniPur®
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
MDL Number: MFCD00011078
CAS Number: 7447-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR