Ethidium bromide 10 mg/mL in aqueous solution, OmniPur®, Millipore®

Supplier: MilliporeSigma
Danger

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

4410-10ML
EM-4410EA 117.89 USD
EM-4410
Ethidium bromide 10 mg/mL in aqueous solution, OmniPur®, Millipore®
Ethidium bromide

For molecular biology. Dark red haze-free liquid. DNase-, RNase-, and protease-free.

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR