EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥97.0%, OmniPur®, Millipore®

Supplier: MilliporeSigma

4100-50GM
EM-4100EA 378.35 USD
EM-4100
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥97.0%, OmniPur®, Millipore®
EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)

For molecular biology. DNase-, RNase-, and protease-free.

Formula: C₁₄H₂₄N₂O₁₀
Melting Pt: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00004291
CAS Number: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR