EDTA (ethylenediamine tetraacetic acid) ≥99.5%, OmniPur® for biotechnology, Millipore®

Supplier: MilliporeSigma
Warning

4005-500GM
EM-4005EA 172.37 USD
EM-4005 EM-4008
EDTA (ethylenediamine tetraacetic acid) ≥99.5%, OmniPur® for biotechnology, Millipore®
EDTA (Ethylenediamine tetraacetic acid)

For molecular biology. DNase-, RNase-, and protease-free.

Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm3 (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR