Dextran sulfate sodium salt solution 50%, OmniPur®, sterile, Calbiochem®

Supplier: MilliporeSigma

3730-100ML
EM-3730EA 557.25 USD
EM-3730
Dextran sulfate sodium salt solution 50%, OmniPur®, sterile, Calbiochem®
Dextran sulphate sodium salt

Caution: Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now

Specification Test Results

DNase None detected
Protease None detected
RNase None detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR