Ammonium sulfate, OmniPur® for biotechnology, Millipore®

Supplier: MilliporeSigma

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

2170-25KG 2150-1KG 2160-5KG
EM-2170EA 931 USD
EM-2170 EM-2150 EM-2160
Ammonium sulfate, OmniPur® for biotechnology, Millipore®
Ammonium sulphate

For molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: >280 °C (dec., lit.)
Density: 1.77 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR