Alanine allyl ester

Supplier: BACHEM AMERICAS INC

4028710.0001 4028710.0005
E-3505.0001BAEA 147.06 USD
E-3505.0001BA E-3505.0005BA
Alanine allyl ester
Alanine allyl ester

Sequence: H-Ala-allyl ester · p-tosylate


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₆H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD02259514
CAS Number: 44812-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR