Glycine

Supplier: BACHEM AMERICAS INC

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

4030676.0500
E-3000.0500BAEA 115.91 USD
E-3000.0500BA
Glycine
Glycine

Sequence: H-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: NH₂CH₂COOH
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR