L-Phenylalanine

Supplier: BACHEM AMERICAS INC

4031148.0250 4031148.1000
E-2230.0250BAEA 113.97 USD
E-2230.0250BA E-2230.1000BA
L-Phenylalanine
L-α-Phenylalanine

Sequence: H-Phe-OH


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR