L(+)-Histidine

Supplier: BACHEM AMERICAS INC

Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine

4030670.0250
E-2010.0250BAEA 175.08 USD
E-2010.0250BA
L(+)-Histidine
L(+)-Histidine

Sequence: H-His-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
EINECS: 200-745-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR