L(+)-Glutamine

Supplier: BACHEM AMERICAS INC

4030662.0100
E-1915.0100BAEA 111.18 USD
E-1915.0100BA
L(+)-Glutamine
L(+)-Glutamine

Sequence: H-Gln-OH


For isoglutamine derivatives please see the family "Glutamic Acid".

Formula: C₅H₁₀N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR