L(+)-Cysteine hydrochloride monohydrate

Supplier: BACHEM AMERICAS INC

4030461.0100
E-1755.0100BAEA 348.03 USD
E-1755.0100BA
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate

Sequence: H-Cys-OH · HCl · H₂O


Due to its high reactivity towards formaldehyde, cysteine should be added as scavenger when cleaving His(Bom)-containing peptides, especially in case of an N-terminal Cys. CAS Number (net): 52-90-4.

Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR