Sulfide, Vacu-vials Instrumental Kit, CHEMetrics
Supplier: CHEMetrics
K-9503
CXK-9503EA 61.57 USD
CXK-9503
Sulfide, Vacu-vials Instrumental Kit, CHEMetrics
Environmental Test Kits
CHEMetrics test kits measure total acid soluble sulfides and employ the methylene blue methodology. Sulfides react with dimethyl-p-phenylenediamine in the presence of ferric chloride to produce methylene blue.
  • Analysis Time: 5 minutes
Sulfides are naturally present in ground waters as a result of leaching from sulfur-containing mineral deposits. Surface waters do not usually contain high sulfide concentrations. Sulfides result from the decomposition of organic matter, from bacterial sulfate reduction under anaerobic conditions and from various chemical processes.
Ordering information: This kit requires the use of a CHEMetrics Direct-Readout Photometer eg V2000 or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.
Delivery information: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Activator Solution, 25 ml sample cup, ampoule blank, and instructions.