Ozone Vacu-vials Instrumental Kit, CHEMetrics
Supplier: CHEMetrics
K-7423
CXK-7423EA 87.49 USD
CXK-7423
Ozone Vacu-vials Instrumental Kit, CHEMetrics
Environmental Test Kits
This kit requires the use of the V-2000 or V-3000 Photometer, a SAM Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell.
  • Analysis time: 1 minute
  • DPD method, 0 to 5.00 ppm
Delivery information: It contains everything needed to perform 30 tests: thirty ampoules, activator Solution, 25 ml sample cup, ampoule blank, and instructions.
Packaging: Ozone Test Kit comes in a cardboard box.