Camco® Erada-Stain® Biological and Chemical Stain Removing Cream, Cambridge
Supplier: Cambridge Diagnostic Products
206
CM206EA 22.88 USD
CM206
Camco® Erada-Stain® Biological and Chemical Stain Removing Cream, Cambridge
Skincare Soaps and Skin Cleansers
This mild cream cleaner and stain remover is effective on most histological dyes, tinctures, general laboratory chemicals, and most other medical stains, including Wright stain
Simply apply and rub gently onto hands for a short time and rinse with water. Cream is biodegradable and phosphate-free. Cream can also be used to clean plastic, glassware, and laboratory equipment.