Sodium acetate trihydrate 99.0-101.0% (by titrimetric analysis) ACS
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Acetic acid sodium salt trihydrate
01258-1KG
01258-2.5KG
01258-1KG
01258-2.5KG
102573-430EA
43.85
USD
102573-430
102573-432
CH01258-1KG
CH01258-2.5KG
Sodium acetate trihydrate 99.0-101.0% (by titrimetric analysis) ACS
Sodium acetate trihydrate
Formula:
H₃CCOONa·3H₂O Melting Pt: ~58 °C |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 |
|
Specification Test Results
Appearance | White crystals |
Melting Point | 58°C (Approx.) |
Heavy Metals (as Pb) | ≤5 ppm |
Anion Chloride (Cl⁻) | ≤0.001% |
Phosphate (PO₄³⁻) | ≤5 ppm |
Sulfate (SO₄²⁻) | ≤0.002% |
Calcium (Ca) | ≤0.005% |
Magnesium (Mg) | ≤0.002% |
Potassium (K) | ≤0.005% |
Iron (Fe) | ≤5 ppm |
pH | 7.5 - 9.2 (5% solution at 25°C) |
Insoluble material | ≤0.005% |
Assay by titration | 99.0-101.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...