Sodium acetate trihydrate 99.0-101.0% (by titrimetric analysis) ACS

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Acetic acid sodium salt trihydrate

01258-1KG 01258-2.5KG 01258-1KG 01258-2.5KG
102573-430EA 43.85 USD
102573-430 102573-432 CH01258-1KG CH01258-2.5KG
Sodium acetate trihydrate 99.0-101.0% (by titrimetric analysis) ACS
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
Melting Pt: ~58 °C
MDL Number: MFCD00071557
CAS Number: 6131-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White crystals
Melting Point 58°C (Approx.)
Heavy Metals (as Pb) ≤5 ppm
Anion Chloride (Cl⁻) ≤0.001%
Phosphate (PO₄³⁻) ≤5 ppm
Sulfate (SO₄²⁻) ≤0.002%
Calcium (Ca) ≤0.005%
Magnesium (Mg) ≤0.002%
Potassium (K) ≤0.005%
Iron (Fe) ≤5 ppm
pH 7.5 - 9.2 (5% solution at 25°C)
Insoluble material ≤0.005%
Assay by titration 99.0-101.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR