L-Methionine 98.5-101.5% (by titrimetric analysis) USP
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
(S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
00221-25KG
00221-100G
00221-250G
00221-25G
00221-1KG
00221-5KG
00221-100G
00221-250G
102553-576EA
2076.45
USD
102553-576
102572-602
102572-604
102614-880
102614-882
102641-466
CH00221-100G
CH00221-250G
L-Methionine 98.5-101.5% (by titrimetric analysis) USP
L-Methionine
Formula:
C₅H₁₁NO₂S MW: 149.21 g/mol Boiling Pt: 278 Melting Pt: 136 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063097 CAS Number: 63-68-3 Merck Index: 13,06004 |
Specification Test Results
Heavy Metals | ≤15 ppm |
Chloride (Cl⁻) | ≤0.05% |
Sulphate (SO₄²⁻) | ≤0.03% |
Iron (Fe) | ≤30 ppm |
pH | 5.6 - 6.1 |
Optical Rotation | [a]D20 = +22.4 to +24.7° |
Infrared Spectrum | Conforms to Structure |
Loss on Drying | ≤0.3% |
Organic Volatile Impurities | To Meet Requirements |
Individual Impurity | ≤0.5% |
Total Impurities | ≤2.0% |
Residue on Ignition | ≤0.4% |
Assay by Titration | 98.5 - 101.5% (Anhydrous Basis) |
Appearance | White to off-white crystalline powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...