EDTA disodium salt dihydrate ≥99%, powder ACS

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

00138-25KG 00138-250G 00138-1KG 00138-2.5KG 00138-10KG 00138-5KG 00138-10KG 00138-1KG 00138-250G 00138-25KG 00138-5KG
102561-422EA 3078.22 USD
102561-422 102572-456 102572-458 102572-460 102572-462 102631-350 CH00138-10KG CH00138-1KG CH00138-250G CH00138-25KG CH00138-5KG
EDTA disodium salt dihydrate ≥99%, powder ACS
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Molecular Formula C10H14N2O8Na2·2H2O
Molecular Weight 372.2
MDL Number MFCD00150037
Storage Temperature Store at RT
Color White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR