EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥99% (by titrimetric analysis), powder

Supplier: Chem-Impex International

00050-100G 00050-10KG 00050-5G 00050-25G 00050-1KG 00050-100G 00050-250G 00050-25G 00050-5G
102586-990EA 102.31 USD
102586-990 102586-994 102614-546 102614-548 102641-460 CH00050-100G CH00050-250G CH00050-25G CH00050-5G
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) ≥99% (by titrimetric analysis), powder
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

A carbodiimide coupling reagent that generates a urea by-product which can be easily removed from the reaction media by extraction with water

Formula: C₈H₁₇N₃·HCl
MDL Number: MFCD00012503
CAS Number: 25952-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Molecular Formula C8H17N3·HCl
Molecular Weight 191.7
MDL Number MFCD00012503
Storage Temperature Store at 0-8°C
Color White to Off-White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR