VWR® Contour™ Touch-Up Paint
Supplier: VWR International
CF-1921-61
CF-1921-61EA 174.69 USD
CF-1921-61
VWR® Contour™ Touch-Up Paint
Sprays
High quality air-dry enamel to match VWR® Contour casework.
  • Paint can be used to touch up VWR® Contour casework
Supplied in a 237 mL (8 oz.) can.