Bench Top Rotating Sample Divider (BTRSD), Rocklabs
Supplier: ROCKLABS (W/T)
300985
CA76047-488EA 30825.81 USD
CA76047-488
Bench Top Rotating Sample Divider (BTRSD), Rocklabs
Mills
The Bench Top Rotating Sample Divider is designed to be placed on a work bench, It can also be used in conjunction with a Bench Top Ring Mill.
  • A representative split of any proportion from 2 to 50%
  • Two representative splits–sample and remainder
  • Sealed bearings
Taking a representative split from a large sample is not as simple as many people believe. Rolling on a mat, multiple grabs or riffle splitting do not produce accurate results. A classic study conducted at Bradford University demonstrated that a rotating sample divider provided the best results.
Ordering information: Manufacturer's 2000 operating hours or one year warranty (whichever comes first), covering all parts (excluding wear parts) and labour if used under normal laboratory conditions. Dust extraction not available.