Sulfuric acid 50% (v/v)

Supplier: VWR
Danger

88442-960
CA71008-974EA 2268.19 USD
CA71008-974
Sulfuric acid 50% (v/v)
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR