Potassium cyanide, Technical Grade

Supplier: VWR
Danger

73290-580
CA71008-088EA 910.97 USD
CA71008-088
Potassium cyanide, Technical Grade
Potassium cyanide
Formula: KCN
MW: 65.12 g/mol
Boiling Pt: 1625 °C (1013 hPa)
Melting Pt: 634 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I
Merck Index: 13,07709

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR