Ethanol denatured ≥90% (v/v), Accusolv™ (denatured with 5% isopropanol and 5% methanol)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

38318-540
CA71007-214CS 1382.6 USD
CA71007-214
Ethanol denatured ≥90% (v/v), Accusolv™ (denatured with 5% isopropanol and 5% methanol)
Ethanol denatured

Suitable for Spectrophotometry

Formula: H₃CCH₂OH
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,III

Order Now

Specification Test Results

UV absorbance (1 cm cell vs H2O) 210 nm 1.000 max.
UV absorbance (1 cm cell vs H2O) 220 nm 0.500 max.
UV absorbance (1 cm cell vs H2O) 230 nm 0.200 max.
UV absorbance (1 cm cell vs H2O) 250 nm 0.050 max.
UV absorbance (1 cm cell vs H2O) 270 nm 0.010 max.
Ethanol 89 - 91% v/v
Methanol 4 - 6 % v/v
Isopropyl Alcohol 4 - 6 % v/v
Water (H2O) 0.1% max.
Titrable acid 0.0005 meq/g max.
Titrable base 0.0002 meq/g max.
Residue after evaporation 5 mg/L max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR