Acetic acid ≥99.7% ACS

Supplier: Anachemia
Danger

6000-4G250C 00598 -461 6000-4H250C
CA71006-424CS 512.95 USD
CA71006-424 CA71006-426 CA11021-244
Acetic acid ≥99.7% ACS
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH3COOH) 99.7% min.
Color (APHA) 10 max.
Dilution test Passes Test
Residue after evaporation 0.001 % max.
Acetic Anhydride 0.01 % max.
Chloride (Cl) 1 ppm max.
Sulfate (SO4) 1 ppm max.
Heavy metals (as Pb) 0.5 ppm max.
Iron (Fe) 0.2 ppm max.
Substances reducing dichromate Passes Test
Substances reducing KMnO4 Passes Test
Titrable base 0.0004 meq/g max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR