LaMotte® Pondwater Tour
Supplier: LaMotte
5418-CN
CA470334-192EA 143.5 USD
CA470334-192
LaMotte® Pondwater Tour
Educational Materials Environmental Educational Materials Environmental Learning Activities
Study water quality of lakes, streams, ponds, and even aquariums.
  • Test for ammonia, dissolved oxygen, nitrate, and pH
  • Individually sealed foil reagent tablets
  • Materials for 50 students
With this activity, students will investigate the natural process and variables that create changes in water quality over a period of time. They will collect samples from any local freshwater source, then use reagent tablets in individually sealed foil strips to test the ammonia, dissolved oxygen, and nitrate contents, as well as the pH of the sample.