Moh's Hardness Collection
Supplier: Avantor
CA470313-110EA 39.39 USD
CA470313-110
Moh's Hardness Collection
Specimens Mineral Specimens Mineral Collections
Our most popular collection.
  • Nine numbered samples
  • Includes streak plate, nail, glass slide, and copper strip
All samples are pure crystalline forms to ensure accurate test results. You can also perform other physical properties tests with the included materials. The set is housed in a compartmented collection box, and an identification list is included.