n-Pentane, Accusolv™, distilled

Supplier: Avantor
Danger

Synonyms: Pentane

CA11020-634CS 1289.52 USD
CA11020-634
n-Pentane, Accusolv™, distilled
n-Pentane

Suitable for HPLC, Spectrophotometry and Pesticide Residue Analysis.

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

UV absorbance (1 cm cell vs H2O) 190 nm 1.000 max.
UV absorbance (1 cm cell vs H2O) 200 nm 0.600 max.
UV absorbance (1 cm cell vs H2O) 250 nm 0.010 max.
UV absorbance (1 cm cell vs H2O) 300 nm 0.005 max.
UV absorbance (1 cm cell vs H2O) 400 nm 0.005 max.
Assay (GC) as n-Pentane 98.0% min.
Assay (GC) as saturated C5 hydrocarbons 99.9% min.
Water (H2O) 0.01% max.
Residue after evaporation 1 mg/l max.
Electron capture GC as heptachlorepoxide 10 ng/l max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR