Wet-Dry Sieve Shaker, Sepor
Supplier: SEPOR CA
040B-001
CA11020-358EA 1038.07 USD
CA11020-358
Wet-Dry Sieve Shaker, Sepor
Shakers and Mixers Sieve Shakers
The Wet-Dry Sieve Shaker is supplied for the purpose of conducting relatively simple size analysis for materials
The sieve shaker uses standard 8 inch diameter laboratory sieve screens and has a capacity for 2 full height 8" diameter sieve screens. The smallest opening sieve screen that should be used with the Wet-Dry Sieve Shaker is 100 mesh.
Ordering information: Contact your Avantor sales representative for more information.