Bead Anvil, Denco Equipment and Machining
Supplier: DENCO EQUIPMENT & MACHINING CO. CA
FA120
CA11019-682EA 88.18 USD
CA11019-682
Bead Anvil, Denco Equipment and Machining
Tool Sets
Ideal for flatenning beads, the bead anvil is constructed from shaft steel with a hardened surface.
Ordering information: Contact your Avantor sales representative for more information.