A.P. Green Fire Assay Crucibles, HarbisonWalker
Supplier: HarbisonWalker
83472 54686 54688 54691 54694 54684 54697 112531 54698
CA11019-246CS 251.46 USD
CA11019-246 11019-256 11019-258 11019-260 11019-262 11019-264 11019-266 80704-052 80704-054
A.P. Green Fire Assay Crucibles, HarbisonWalker
Crucibles
A. P. Green Fire Assay Crucibles have several features which make them ideal for mining assays.
  • Withstand thermal shock
  • Allow for more heats per crucible
  • Multiple size options available
Their scientific mix of material batches, proper pressing with accurate dyes, and the correct graduating temperatures at which they are burned, provide the highest degree of mechanical strength to withstand severe thermal shock. The high quality materials used in their manufacture, coupled with carefully managed production, ensures more heats per crucible, thus providing more value. Combine all these features with the convenience of several available sizes and shapes, and A.P. Green Fire Assay Crucibles become the perfect choice for mining applications.
Ordering information: Multiple size options are available. Contact your Avantor sales Representative for more information.