Hollow Cathode Lamps to Suit All AAS Systems, Photron
Supplier: PHOTRON PTY LTD.
P802 P804 P806 P809 P812 P820 P831 P839 P840 P841 P849 P852 P855 P870 P871 P873P873 P803 P808 P813 P814 P821 P826 P828 P832 P833 P834 P836 P851 P821C P851C P867 P872 P803C P524 P527
CA11017-770EA 586.14 USD
CA11017-770 CA11017-774 CA11017-778 CA11017-784 CA11017-790 CA11017-806 CA11017-828 CA11017-844 CA11017-846 CA11017-848 CA11017-864 CA11017-870 CA11017-876 CA11017-978 CA11017-980 CA11017-984 11017-772 11017-782 11017-792 11017-794 11017-808 11017-818 11017-822 11017-830 11017-832 11017-834 11017-838 11017-868 11017-908 11017-968 11017-972 11017-982 11017-992 11018-522 11018-528
Hollow Cathode Lamps to Suit All AAS Systems, Photron
Lights Hollow Cathode Lamps
Hollow cathode lamps produced by Photron are designed and manufactured to achieve all of the following fundamental requirements for a spectral line source: Intense emission of resonance (ground state derived) lines. Narrow line width, for maximum sensitivity and linearity. Minimal spectral interference from continuum emission, present in the cathode. Rapid warm up to produce stable long term light emission. Noise free operation. Long running life and a 5 year shelf life. Cathode materials and geometry achieve a combination of spectral purity and a suitable sputtering rate. (A high sputtering rate will give high intensity at the expense of lamp life, sensitivity and linearity).
Photron's P800 (37mm or 1.5” Dia.) series is designed for use in AAS systems manufactured by GBC Scientific, Varian, Analytic Jena, Shimadzu, and other makes
Photron's P500 Multi Element (37mm or 1.5” Dia.) series is designed for direct use in AAS systems manufactured by GBC Scientific, Varian, Analytic Jena, Shimadzu, Hitachi, Thermo Jarrel Ash, Unicam, and other makes.
SMITH-HIEFTJE background correction. Photron lamps have been tested and found to work as well as lamps sold specifically for this technique, however due to the high energy pulse used, some elements will be consumed at a higher rate reducing lamp life.
Ordering information: Contact your Avantor sales representative for more information. Wavelength listed is the primary wavelength.