Furnace Shovel, Steel, Webbers Welding
Supplier: Webber's Welding
FA129
CA11017-664EA 449.86 USD
CA11017-664
Furnace Shovel, Steel, Webbers Welding
Scrapers
The furnace shovel is ideal for either catching scrapings when cleaning the furnace or adding bone ash
The handle is constructed of durable 0.6cm steel rod with a 25.4cm wide x 20.3cm deep pan size for maximum shoveling efficiency
Ordering information: Contact your Avantor sales representative for more information.