Cupel Tongs, Heavy-Duty Steel, Webber's Welding
Supplier: Webber's Welding
FA117
CA11017-658EA 449.86 USD
CA11017-658
Cupel Tongs, Heavy-Duty Steel, Webber's Welding
Tongs
The Cupel Tongs are specially designed for heavy-duty applications with user safety in mind.
  • Heavy-duty steel construction provides durable and dependable service
  • Longer length increases user safety, by distancing the user from the oven or furnace