Tris(carbobenzoxy)-L-arginine

Supplier: BACHEM AMERICAS INC

Synonyms: Nα,Nδ,Nomega-Tris(benzyloxycarbonyl)-L-arginine, Tri-Cbz-L-arginine, Z-Arg(Z)2-OH, Z-Arg(Cbz)₂-OH

4000138.0005 4000138.0025 4000138.0100
C-2975.0005BAEA 147.06 USD
C-2975.0005BA C-2975.0025BA C-2975.0100BA
Tris(carbobenzoxy)-L-arginine
Tris(carbobenzoxy)-L-arginine

Sequence: Z-Arg(Z)₂-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₀H₃₂N₄O₈
MW: 576.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038693
CAS Number: 14611-34-8
EINECS: 238-647-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR