1,7-Octadiyne ≥98%

Supplier: BeanTown Chemical

Synonyms: Octa-1,7-diyne

225945-10G 225945-50G
BT225945-10GEA 81.43 USD
BT225945-10G BT225945-50G
1,7-Octadiyne ≥98%
1,7-Octadiyne

CAS: 871-84-1; EC No: 212-815-0; MDL No: MFCD00008580 UN No: UN3295; Haz Class: 3; Packing Group: III Liquid; Linear Formula: HC≡C(CH2)4C≡CH; Molecular Formula: C8H10; MW: 106.17 Boiling Point: 135-136°; Flash point: 23°C (73°F) Density (g/mL): 0.8; Refractive Index: 1.446


  • Refractive index: 1.446
  • Liquid

Formula: C₈H₁₀
Boiling Pt: 135…136 °C
Density: 0.8 g/mL
Flash Pt: 23 °C (73 °F)
MDL Number: MFCD00008580
CAS Number: 871-84-1
EINECS: 212-815-0
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR