Graphite (natural) ≥99.9% (trace metals basis), powder -20+84 mesh

Supplier: BeanTown Chemical

225915-1KG 225915-250G
BT225915-1KGEA 75 USD
BT225915-1KG BT225915-250G
Graphite (natural) ≥99.9% (trace metals basis), powder -20+84 mesh
Graphite

CAS: 7782-42-5; EC No: 231-955-3; MDL No: MFCD00144065 Powder; Molecular Formula: C; MW: 12.011


  • Powder

Formula: C
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR