Selenium ≥99.5% (trace metals basis), powder -325 mesh

Supplier: BeanTown Chemical

225745-250G 225745-50G
BT225745-250GEA 357 USD
BT225745-250G BT225745-50G
Selenium ≥99.5% (trace metals basis), powder -325 mesh
Selenium

CAS: 7782-49-2; EC No: 231-957-4; MDL No: MFCD00134090; RTECS: VS7700000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: Se; MW: 78.96 Melting Point: 217°; Boiling Point: 684.9° Density (g/mL): 4.81


  • RTECS: VS7700000
  • Powder

Formula: Se
Boiling Pt: 684.9 °C
Melting Pt: 217 °C
Density: 4.81 g/mL
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3077
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR