Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Supplier: BeanTown Chemical
225725-100G
225725-2.5KG
225725-500G
BT225725-100GEA
30
USD
BT225725-100G
BT225725-2.5KG
BT225725-500G
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
CAS: 25805-17-8; MDL No: MFCD00134377
Solid; Linear Formula: [-N(COC2H5)CH2CH2-]n ; MW ≈200,000
Density (g/mL): 1.14
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
(C₅H₉NO)x Density: 1.14 g/mL |
CAS Number:
25805-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...