Barium stearate
Supplier: BeanTown Chemical
Synonyms:
Stearic acid barium salt
225705-25G
BT225705-25GEA
31
USD
BT225705-25G
Barium stearate
Barium stearate
CAS: 6865-35-6; EC No: 229-966-3; MDL No: MFCD00050711; RTECS: WI2840000
Solid; Linear Formula: Na(O2C18H35)2; Molecular Formula: C36H70BaO4; MW: 704.28
- RTECS: WI2840000
- Solid
Formula:
C₃₆H₇₀BaO₄ |
MDL Number:
MFCD00050711 CAS Number: 6865-35-6 EINECS: 229-966-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...