(S)-tert-Butyl-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate

Supplier: BeanTown Chemical

Synonyms: tert-Butyl-(S)-(3-amino-2-oxo-2,3,4,5-tetrahydrobenzo[b]azepin-1-yl) acetate, Benazepril Related Compound F, (S)-tert-Butyl 2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate

225670-100G 225670-25G 225670-5G
BT225670-100GEA 758 USD
BT225670-100G BT225670-25G BT225670-5G
(S)-tert-Butyl-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate
(S)-tert-Butyl-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate

CAS: 109010-60-8; MDL No: MFCD00895734; RTECS: CX7065000 Solid; Molecular Formula: C16H22N2O3; MW: 290.36


  • RTECS: CX7065000
  • Solid

Formula: C₁₆H₂₂N₂O₃
MDL Number: MFCD03426719
CAS Number: 109010-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR