Zinc selenide, crystals optical disc, polished both sides, Length 13 mm, Width 2 mm

Supplier: BeanTown Chemical

Synonyms: Zinc (II) selenide

225660-1EACH
BT225660-1EACHEA 274 USD
BT225660-1EACH
Zinc selenide, crystals optical disc, polished both sides, Length 13 mm, Width 2 mm
Zinc selenide

CAS: 1315-09-9; EC No: 215-259-7 Optic Disc

Formula: SeZn
MW: 144.35 g/mol
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR